This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EIF4A1 SIRNA Assays ›
Gene Symbol:
Eif4a1
Gene Name:
eukaryotic translation initiation factor 4A1
Gene Aliases:
BM-010, Ddx2a, Eif4
Chromosome Location:
Chr.11: 69666936 - 69672423 on Build GRCm38
UniGene ID:
Mm.26680
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s65417

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Eif4a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144958.4 NP_659207.1 10 1189
GenBank mRNA AK010644.1 - 10 1090
AK077429.1 - 10 1082
AK088124.1 - 6 2478
AK135752.1 - 10 1082
AK135994.1 - 10 1083
AK144850.1 - 10 1082
AK145375.1 - 10 1084
AK145508.1 - 10 1084
AK146424.1 - 10 1084
AK146464.1 - 10 1082
AK152210.1 - 7 865
AK152694.1 - 10 1084
AK159643.1 - 10 1084
AK162017.1 - 9 1038
AK166527.1 - 10 1082
AK166572.1 - 10 1084
AK166674.1 - 10 1084
AK167003.1 - 10 1082
AK167083.1 - 10 1082
AK167196.1 - 10 1084
AK167205.1 - 10 1081
AK167906.1 - 9 1154
AK167988.1 - 10 1082
AK168697.1 - 10 1083
AK168780.1 - 10 1082
AK169023.1 - 10 1082
AK169030.1 - 10 1075
AK169057.1 - 10 1082
AK169156.1 - 10 1082
AK169279.1 - 10 1082
AK190420.1 - 2 113
AK196626.1 - 2 113
AK204226.1 - 2 113
AK212197.1 - 2 113
BC002266.1 - 1 53
BC012282.1 - 10 1071
BC049915.1 - 10 1072 5068709
BC099392.1 - 9 1065
X03039.1 - 10 1079
X03040.1 - 9 1082

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_199372, XM_006246624, FQ225967, FQ212737, FQ233795, and others.

Gene Ontology Categories:

Function(s) Process(es)

translation
translational initiation
regulation of gene expression
RNA secondary structure unwinding
nucleotide binding
nucleic acid binding
RNA binding
double-stranded RNA binding
translation initiation factor activity
ATP-dependent RNA helicase activity
helicase activity
protein binding
ATP binding
hydrolase activity
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0