This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EHF SIRNA Assays ›
Gene Symbol:
Ehf
Gene Name:
ets homologous factor
Gene Aliases:
9030625L19Rik, AU019492
Chromosome Location:
Chr.2: 103263431 - 103303275 on Build GRCm38
UniGene ID:
Mm.10724
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s65395

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ehf
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007914.3 NP_031940.1 5, 6 625
GenBank mRNA AF035527.1 - 4, 5 623
AK028651.1 - 5, 6 596
BC005520.1 - 6, 7 542 3494644
BC006789.1 - 5, 6 607 3589528

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106493, XM_006234641, XM_006234639, BC166742.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0