This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KLK1B26 SIRNA Assays ›
Gene Symbol:
Klk1b26 Egfbp2
Gene Name:
kallikrein 1-related petidase b26
epidermal growth factor binding protein type B
Gene Aliases:
Egfbp2, Klk26, PRECE-2, mGK-26, mGK26
Egfbp-2, Klk1b26, mGk-13
Chromosome Location:
Chr.7: 44007953 - 44016969 on Build GRCm38
Chromosome Location:
-
UniGene ID:
Mm.439663
Mm.439663
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s65368

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Klk1b26
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010644.2 NP_034774.1 4 609
GenBank mRNA AK075752.1 - 2 165
AK075766.1 - 2 165
BC024688.1 - 4 583 4167696
BC138704.1 - 4 591 8861726
K01831.1 - 4 609
X63327.1 - 4 588
Gene Symbol: Egfbp2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010115.6 NP_034245.3 4 591
GenBank mRNA AB016032.1 - 4 567
X58628.1 - 4 591

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

brain renin-angiotensin system
proteolysis
zymogen activation
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header