This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DNAH8 SIRNA Assays ›
Gene Symbol:
Dnah8
Gene Name:
dynein, axonemal, heavy chain 8
Gene Aliases:
ATPase, Dnahc8, Hst6.7b, P1-Loop
Chromosome Location:
Chr.17: 30626936 - 30875762 on Build GRCm38
UniGene ID:
Mm.426017
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s65042

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dnah8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013811.3 NP_038839.2 1654
GenBank mRNA AF356520.1 - 1654
AF356521.1 - 1654
AF356522.1 - 1664
AF356523.1 - 1664
AF363577.1 - 1664
AK161258.1 - 1656

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

microtubule-based movement
biological_process
nucleotide binding
molecular_function
motor activity
microtubule motor activity
ATP binding
ATPase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0