This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CRYBB3 SIRNA Assays ›
Gene Symbol:
Crybb3
Gene Name:
crystallin, beta B3
Gene Aliases:
AI852419
Chromosome Location:
Chr.5: 113075839 - 113082233 on Build GRCm38
UniGene ID:
Mm.378899
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s64471

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Crybb3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159650.1 NP_001153122.1 5 446
NM_021352.3 NP_067327.1 5 516
GenBank mRNA AJ272229.2 - 4 500
AK053552.1 - 3 830
BC031442.1 - 4 507 4501690
BC058522.1 - 4 425 6824229
CJ046471.1 - 5 517

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_031690, XM_006249572, X05899, AF287304.

Gene Ontology Categories:

Process(es)

structural constituent of eye lens

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0