This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CR2 SIRNA Assays ›
Gene Symbol:
Cr2
Gene Name:
complement receptor 2
Gene Aliases:
C3DR, CD21, CD35, Cr-1, Cr-2, Cr1
Chromosome Location:
Chr.1: 195136811 - 195177190 on Build GRCm38
UniGene ID:
Mm.235387
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s64355

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cr2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007758.2 NP_031784.1 8 1565
GenBank mRNA AK003072.2 - 8 1565
BC109344.1 - 8 1498 40054528
M29281.1 - 3 569
M35684.1 - 7 1433
M61132.1 - 7 1400

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

immune system process
complement receptor mediated signaling pathway
complement activation, classical pathway
B cell differentiation
B cell proliferation
B cell activation
innate immune response
complement binding
DNA binding
receptor activity
complement receptor activity
protein binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0