This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other COL5A2 SIRNA Assays ›
Gene Symbol:
Col5a2
Gene Name:
collagen, type V, alpha 2
Gene Aliases:
1110014L14Rik, D230017N05
Chromosome Location:
Chr.1: 45374331 - 45503282 on Build GRCm38
UniGene ID:
Mm.10299
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s64240

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Col5a2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007737.2 NP_031763.2 53 4587
GenBank mRNA AK084285.1 - 15 1404
AK084538.1 - 12 1371
AK084803.1 - 53 4226
AK132413.1 - 53 4369
AK139130.1 - 53 4162
AK147220.1 - 53 4369
AK147328.1 - 53 4588
AK151929.1 - 53 4371
AK160008.1 - 53 4161
BC043696.1 - 27 2402
BC055077.1 - 53 4338 30059991
L02918.1 - 53 4219

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
collagen fibril organization
skin development
eye morphogenesis
cellular response to amino acid stimulus
negative regulation of endodermal cell differentiation
extracellular matrix structural constituent
SMAD binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header