This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other COL4A5 SIRNA Assays ›
Gene Symbol:
Col4a5
Gene Name:
collagen, type IV, alpha 5
Gene Aliases:
-
Chromosome Location:
Chr.X: 141475369 - 141689236 on Build GRCm38
UniGene ID:
Mm.286892
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s64236

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Col4a5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163155.1 NP_001156627.1 2 438
NM_007736.4 NP_031762.2 3 266
GenBank mRNA AB041350.1 - 2 296
AK133179.1 - 3 267
BC082788.1 - 2 277 5343488
CN668672.1 - 2 383

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008773475, XM_006257312, XM_343778, XM_006257313.

Gene Ontology Categories:

Function(s)

neuromuscular junction development

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0