This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CIRBP SIRNA Assays ›
Gene Symbol:
Cirbp
Gene Name:
cold inducible RNA binding protein
Gene Aliases:
Cirp, R74941
Chromosome Location:
Chr.10: 80165990 - 80172691 on Build GRCm38
UniGene ID:
Mm.17898
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s63987

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cirbp
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007705.2 NP_031731.1 3 248
GenBank mRNA AK014655.1 - 3 251
AK132584.1 - 3 251
AK188303.1 - 2 174
BC075699.1 - 3 238 30656047
D78135.1 - 3 248

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_031147, XM_006241015, XM_006241012, XM_006241014, XM_006241013, and others.

Gene Ontology Categories:

Function(s) Process(es)

response to UV
negative regulation of translation
stress granule assembly
positive regulation of translation
mRNA stabilization
nucleotide binding
nucleic acid binding
RNA binding
mRNA 3'-UTR binding
translation repressor activity
poly(A) RNA binding
small ribosomal subunit rRNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0