This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CDK1 SIRNA Assays ›
Gene Symbol:
Cdk1
Gene Name:
cyclin-dependent kinase 1
Gene Aliases:
Cdc2, Cdc2a, p34<CDC2>
Chromosome Location:
Chr.10: 69336635 - 69352912 on Build GRCm38
UniGene ID:
Mm.281367
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s63733

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cdk1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007659.3 NP_031685.2 3 280
GenBank mRNA AK030231.1 - 3 281
AK051859.1 - 3 307
AK135516.1 - 3 284
AK168054.1 - 3 281
BC005614.1 - 2 173
BC024396.1 - 3 258 4977366
M38724.1 - 2 201
U58633.1 - 3 284
X16461.1 - 2 201

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_019296, XM_006256353, FQ227059, BC091549, FQ219810, and others.

Gene Ontology Categories:

Function(s) Process(es)

protein complex assembly
protein phosphorylation
apoptotic process
cell cycle
mitotic nuclear division
mitotic G2 DNA damage checkpoint
cell aging
response to organonitrogen compound
positive regulation of gene expression
phosphorylation
histone phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
chromosome condensation
organ regeneration
positive regulation of protein import into nucleus, translocation
protein localization to kinetochore
negative regulation of apoptotic process
response to ethanol
positive regulation of DNA replication
positive regulation of mitotic cell cycle
cell division
positive regulation of cardiac muscle cell proliferation
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
cyclin-dependent protein serine/threonine kinase activity
protein binding
ATP binding
RNA polymerase II carboxy-terminal domain kinase activity
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
cyclin binding
Hsp70 protein binding
histone kinase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0