This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BARX2 SIRNA Assays ›
Gene Symbol:
Barx2
Gene Name:
BarH-like homeobox 2
Gene Aliases:
2310006E12Rik, Barx2b
Chromosome Location:
Chr.9: 31846044 - 31913285 on Build GRCm38
UniGene ID:
Mm.261044
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s62873

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Barx2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013800.2 NP_038828.2 4 806
GenBank mRNA BC012684.1 - 4 797 3980774
L77900.1 - 4 604

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: BC111572.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
cartilage condensation
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
myotube differentiation
catagen
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0