This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other APBB1 SIRNA Assays ›
Gene Symbol:
Apbb1
Gene Name:
amyloid beta (A4) precursor protein-binding, family B, member 1
Gene Aliases:
Fe65, Rir
Chromosome Location:
Chr.7: 105558465 - 105581653 on Build GRCm38
UniGene ID:
Mm.38469
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s62438

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Apbb1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253885.1 NP_001240814.1 10 1787
NM_001253886.1 NP_001240815.1 9 1781
NM_001253887.1 NP_001240816.1 10 787
NM_009685.3 NP_033815.1 9 1595
GenBank mRNA AF206720.1 - 9 1504
AK030748.1 - 9 1478
AK075730.1 - 10 767
AK083830.1 - 10 1517
AK144340.1 - 10 1788
AK164140.1 - 10 1512
BC022732.1 - 3 162
BC036956.1 - 9 1425
BC048395.1 - 8 933 6392016

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_080478, XM_006229935, XM_006229941, XM_006229940, XM_008759717, and others.

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
double-strand break repair
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
cellular response to DNA damage stimulus
cell cycle arrest
axon guidance
visual learning
chromatin modification
actin filament-based movement
extracellular matrix organization
negative regulation of cell growth
positive regulation of apoptotic process
histone H4 acetylation
negative regulation of neuron differentiation
positive regulation of DNA repair
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein secretion
negative regulation of thymidylate synthase biosynthetic process
protein stabilization
beta-amyloid binding
chromatin binding
protein binding
transcription factor binding
protein complex binding
histone acetyltransferase binding
histone binding
proline-rich region binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0