This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ALPL SIRNA Assays ›
Gene Symbol:
Alpl
Gene Name:
alkaline phosphatase, liver/bone/kidney
Gene Aliases:
APTNAP, Akp-2, Akp2, TNAP, TNSALP
Chromosome Location:
Chr.4: 137741731 - 137796384 on Build GRCm38
UniGene ID:
Mm.288186
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s62207

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Alpl
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287172.1 NP_001274101.1 6 882
NM_001287176.1 NP_001274105.1 6 1005
NM_007431.3 NP_031457.2 6 857
GenBank mRNA AB473959.1 - 5 621
AK076459.1 - 6 792
AK161780.1 - 6 793
AK164002.1 - 6 850
AK166732.1 - 6 791
AK167023.1 - 6 792
AK167481.1 - 6 792
BC065175.1 - 6 780 6807509
J02980.1 - 6 796
X13409.1 - 6 740

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
endochondral ossification
developmental process involved in reproduction
metabolic process
dephosphorylation
response to antibiotic
response to glucocorticoid
cellular response to organic cyclic compound
catalytic activity
alkaline phosphatase activity
protein binding
pyrophosphatase activity
hydrolase activity
phosphatase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0