This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ABCG1 SIRNA Assays ›
Gene Symbol:
Abcg1
Gene Name:
ATP-binding cassette, sub-family G (WHITE), member 1
Gene Aliases:
AW413978, Abc8, White
Chromosome Location:
Chr.17: 31057694 - 31117984 on Build GRCm38
UniGene ID:
Mm.15691
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s61797

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Abcg1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009593.2 NP_033723.1 9 1213
GenBank mRNA AF323659.1 - 9 1209
BC119471.2 - 9 1161 40048166
CV558133.1 - 1 24
U34920.1 - 9 1171
Z48745.1 - 9 1200

Back To Top

More Information


Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transport
lipid transport
cholesterol metabolic process
response to organic substance
negative regulation of macrophage derived foam cell differentiation
regulation of cholesterol esterification
positive regulation of cholesterol efflux
negative regulation of lipid storage
cholesterol transport
intracellular cholesterol transport
cholesterol efflux
phospholipid efflux
response to lipid
low-density lipoprotein particle remodeling
high-density lipoprotein particle remodeling
glycoprotein transport
cholesterol homeostasis
amyloid precursor protein catabolic process
reverse cholesterol transport
positive regulation of cholesterol biosynthetic process
transmembrane transport
phospholipid homeostasis
response to high density lipoprotein particle
toxin transport
nucleotide binding
ATP binding
phospholipid transporter activity
ATPase activity
cholesterol transporter activity
toxin transporter activity
sterol-transporting ATPase activity
glycoprotein transporter activity
protein homodimerization activity
ADP binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0