siRNA Details
| Gene Symbol: LINC01530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034159.1 | 1 | 163 | |||
| GenBank mRNA | AK054965.1 | - | 1 | 160 | ||
| CA867516.1 | - | 1 | 135 | |||
| DA060240.1 | - | 1 | 160 | |||
| DB478715.1 | - | 1 | 163 | |||

LINC01530| Gene Symbol: LINC01530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034159.1 | 1 | 163 | |||
| GenBank mRNA | AK054965.1 | - | 1 | 160 | ||
| CA867516.1 | - | 1 | 135 | |||
| DA060240.1 | - | 1 | 160 | |||
| DB478715.1 | - | 1 | 163 | |||