This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GPR4 SIRNA Assays ›
Gene Symbol:
GPR4
Gene Name:
G protein-coupled receptor 4
Gene Aliases:
-
Chromosome Location:
Chr.19: 45589764 - 45602208 on Build GRCh38
UniGene ID:
Hs.17170
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s6001

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GPR4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005282.2 NP_005273.1 2 1431
XM_017026607.1 XP_016882096.1 2 2019
XM_017026608.1 XP_016882097.1 2 1340
GenBank mRNA AK290698.1 - 2 1384
BC038450.1 - 2 1419
BC067535.1 - 1 790 6971751
BC067536.1 - 1 790 6971752
EU432116.1 - 1 486

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
response to acidic pH
negative regulation of angiogenesis
angiogenesis involved in wound healing
glomerular mesangial cell development
G-protein coupled receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0