This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KRTAP1-1 SIRNA Assays ›
Gene Symbol:
KRTAP1-1
Gene Name:
keratin associated protein 1-1
Gene Aliases:
HB2A, KAP1.1, KAP1.1A, KAP1.1B, KAP1.6, KAP1.7, KRTAP1.1, KRTAP1A, hKAP1.7
Chromosome Location:
Chr.17: 41040559 - 41041461 on Build GRCh38
UniGene ID:
Hs.247934
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s59179

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KRTAP1-1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030967.2 NP_112229.1 1 839
GenBank mRNA AB052868.1 - 2 701
AB052934.1 - 1 839
AB055057.1 - 2 514
AJ406926.1 - 1 808

Back To Top

More Information


Gene Ontology Categories:

Function(s)

biological_process

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0