This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TAF9B SIRNA Assays ›
Gene Symbol:
TAF9B
Gene Name:
TATA-box binding protein associated factor 9b
Gene Aliases:
DN-7, DN7, TAF9L, TAFII31L, TFIID-31
Chromosome Location:
Chr.X: 78129748 - 78139682 on Build GRCh38
UniGene ID:
Hs.592248
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s58378

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TAF9B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015975.4 NP_057059.2 2 176
GenBank mRNA AF220509.1 - 2 127
BC009566.1 - 2 127 3996687
BC010350.1 - 2 141 4276335
BC071649.1 - 2 113 5246715
BC146952.1 - 2 157 9056880
BC146960.1 - 2 157 9056888
BI544376.1 - 2 204

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
positive regulation of cell growth
negative regulation of apoptotic process
protein stabilization
regulation of signal transduction by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
transcription corepressor activity
protein binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0