This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PTMA SIRNA Assays ›
Gene Symbol:
PTMA LOC728026
Gene Name:
prothymosin, alpha
prothymosin alpha-like
Gene Aliases:
TMSA
-
Chromosome Location:
Chr.2: 231708525 - 231713541 on Build GRCh38
Chromosome Location:
Chr.9: 94347222 - 94348219 on Build GRCh38
UniGene ID:
Hs.459927
-
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID s58175

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PTMA
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099285.1 NP_001092755.1 3 331
NM_002823.4 NP_002814.3 3 328
XM_011511518.1 XP_011509820.1 3 1061
XM_011511519.1 XP_011509821.1 3 1058
XM_011511520.1 XP_011509822.1 3 960
XM_011511521.1 XP_011509823.1 3 957
GenBank mRNA AF348514.1 - 3 146
AF452640.1 - 3 628
AK223605.1 - 3 323
AK314106.1 - 3 323
AY169282.1 - 3 146
BC003510.1 - 3 311 3610808
BC022433.1 - 3 327 4277490
BC034921.1 - 3 319 4335961
BC051265.1 - 3 311 3832454
BC066905.1 - 3 249 4830132
BC070480.1 - 3 313 6205582
BC071647.1 - 3 306 5112610
BC071879.1 - 3 308 6425103
BC098420.1 - 3 313 5017164
BM839980.1 - 3 328
BX647606.1 - 1 996
M14483.1 - 3 278
M14630.1 - 3 326
M14794.1 - 3 214
M26708.1 - 3 301
Gene Symbol: LOC728026
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017015408.1 XP_016870897.1 1 802

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_021740, XM_003751278, XM_008769073, FQ227835, FQ225103, and others.
This siRNA also targets the following transcripts in Mouse: NM_008972, AK152032, AK142076, AK084218, BC081453, and others.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header