This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM217 SIRNA Assays ›
Gene Symbol:
TMEM217
Gene Name:
transmembrane protein 217
Gene Aliases:
C6orf128, dJ355M6.2
Chromosome Location:
Chr.6: 37212178 - 37259034 on Build GRCh38
UniGene ID:
Hs.520101
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s57766

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM217
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286401.1 NP_001273330.1 3 1603
NM_145316.3 NP_660359.2 4 1170
NR_104442.1 3 1072
NR_104443.1 2 987
XM_011514367.2 XP_011512669.1 4 1097
XM_011514369.2 XP_011512671.1 2 498
XM_011514370.2 XP_011512672.1 2 488
GenBank mRNA AK098666.1 - 4 1129
BC026012.1 - 3 1042 5269554
BC029657.1 - 2 468 5167418

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0