This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FBXW10 SIRNA Assays ›
Gene Symbol:
FBXW10 CDRT1
Gene Name:
F-box and WD repeat domain containing 10
CMT1A duplicated region transcript 1
Gene Aliases:
Fbw10, HREP, SM25H2, SM2SH2
C170RF1, C17ORF1, C17ORF1A, FBXW10B, FBXW10P1, HREP, SM25H2
Chromosome Location:
Chr.17: 18744013 - 18779349 on Build GRCh38
Chromosome Location:
Chr.17: 15565482 - 15619704 on Build GRCh38
UniGene ID:
Hs.592128
Hs.548021
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID s57472

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FBXW10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267585.1 NP_001254514.1 3 1007
NM_001267586.1 NP_001254515.1 3 1007
NR_051988.1 3 1653
XM_011523625.1 XP_011521927.1 3 1073
XM_011523626.2 XP_011521928.1 3 1073
XM_017024020.1 XP_016879509.1 3 1073
XM_017024021.1 XP_016879510.1 3 1073
GenBank mRNA AK310383.1 - 3 1592
AL136822.1 - 3 945
AY729024.1 - 3 775
BC028364.2 - 3 994 4827344
Gene Symbol: CDRT1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282540.1 NP_001269469.1 3 967
NM_006382.3 NP_006373.2 3 967

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0