This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF222 SIRNA Assays ›
Gene Symbol:
RNF222
Gene Name:
ring finger protein 222
Gene Aliases:
-
Chromosome Location:
Chr.17: 8390704 - 8397835 on Build GRCh38
UniGene ID:
Hs.526550
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s55493

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RNF222
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146684.2 NP_001140156.1 3 517
XM_011523978.2 XP_011522280.1 3 494
XM_011523979.2 XP_011522281.1 3 602
XM_011523980.2 XP_011522282.1 2 386
XM_011523981.2 XP_011522283.1 2 330

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
negative regulation of protein sumoylation
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of transcription from RNA polymerase II promoter
transcription factor binding
zinc ion binding
SUMO polymer binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header