This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PGA5 SIRNA Assays ›
Gene Symbol:
PGA5 PGA3 PGA4
Gene Name:
pepsinogen 5, group I (pepsinogen A)
pepsinogen 3, group I (pepsinogen A)
pepsinogen 4, group I (pepsinogen A)
Gene Aliases:
Pg5
-
-
Chromosome Location:
Chr.11: 61241176 - 61251444 on Build GRCh38
Chromosome Location:
Chr.11: 61203513 - 61212869 on Build GRCh38
Chromosome Location:
Chr.11: 61222349 - 61231695 on Build GRCh38
UniGene ID:
Hs.432854
Hs.601055
Hs.661883
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
siRNA ID s55464

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PGA5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014224.4 NP_055039.1 9 1278
GenBank mRNA BC029055.1 - 9 1257 5184316
BC146999.1 - 9 1278 9056927
BC171889.1 - 9 1278 9054543
BC171897.1 - 9 1278 9054551
Gene Symbol: PGA3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079807.3 NP_001073275.1 9 1279
GenBank mRNA AK125628.1 - 9 1293
AK225669.1 - 9 1277
AK225674.1 - 9 1305
AK225679.1 - 9 1277
AK301293.1 - 7 880
AL832946.1 - 1 2798
BC171815.1 - 9 1273 9054469
Gene Symbol: PGA4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079808.3 NP_001073276.1 9 1279
GenBank mRNA AK291864.1 - 9 1277
AK301305.1 - 6 751
BC150659.1 - 9 1273 9057030
BC171808.1 - 9 1273 9054462
BC171814.1 - 9 1273 9054468
BC171910.1 - 9 1273 9054564
BC171920.1 - 9 1273 9054574

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
digestion
protein catabolic process
cellular protein metabolic process
aspartic-type endopeptidase activity
peptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0