This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ERICH2 SIRNA Assays ›
Gene Symbol:
ERICH2
Gene Name:
glutamate rich 2
Gene Aliases:
-
Chromosome Location:
Chr.2: 170770042 - 170798971 on Build GRCh38
UniGene ID:
Hs.443729
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s55400

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ERICH2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290030.1 NP_001276959.1 2, 3 530
NM_001290031.1 NP_001276960.1 2, 3 530
NR_110644.1 3, 4 837
XM_011510998.2 XP_011509300.1 4, 5 662
XM_011510999.2 XP_011509301.1 3, 4 329
XM_011511001.2 XP_011509303.1 2, 3 1008
XM_017003875.1 XP_016859364.1 3, 4 1031
XM_017003876.1 XP_016859365.1 2, 3 46
XM_017003877.1 XP_016859366.1 3, 4 329
GenBank mRNA AA401652.1 - 2, 3 81
BQ269226.1 - 3, 4 406

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0