This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CT45A3 SIRNA Assays ›
Gene Symbol:
CT45A3 CT45A1 CT45A2 CT45A5 CT45A6 CT45A7 CT45A8
Gene Name:
cancer/testis antigen family 45, member A3
cancer/testis antigen family 45, member A1
cancer/testis antigen family 45, member A2
cancer/testis antigen family 45, member A5
cancer/testis antigen family 45, member A6
cancer/testis antigen family 45, member A7
cancer/testis antigen family 45, member A8
Gene Aliases:
CT45-3, CT45-4, CT45.3, CT45.4, CT45A4
CT45, CT45-1, CT45.1
CT45-2, CT45.2
CT45.5, CT455
CT45-6, CT45.6
-
-
Chromosome Location:
Chr.X: 135760067 - 135768222 on Build GRCh38
Chromosome Location:
Chr.X: 135708398 - 135723318 on Build GRCh38
Chromosome Location:
Chr.X: 135811979 - 135820062 on Build GRCh38
Chromosome Location:
Chr.X: 135777130 - 135785512 on Build GRCh38
Chromosome Location:
Chr.X: 135794687 - 135802755 on Build GRCh38
Chromosome Location:
Chr.X: 135829229 - 135837317 on Build GRCh38
Chromosome Location:
Chr.X: 135846497 - 135854588 on Build GRCh38
UniGene ID:
Hs.535080
Hs.460937
Hs.535081
Hs.535080
Hs.676560
Hs.535080
Hs.568346
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
-
-
-
-
siRNA ID s54381

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CT45A3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017435.1 NP_001017435.1 3 622
XM_006724758.3 XP_006724821.1 3 539
XM_011531340.2 XP_011529642.1 3 498
GenBank mRNA AK098689.1 - 3 467
AY743711.1 - 3 622
Gene Symbol: CT45A1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017417.2 NP_001017417.1 3 621
XM_005278141.3 XP_005278198.1 3 470
XM_011531352.1 XP_011529654.1 2 391
GenBank mRNA AY743709.1 - 3 622
BC140913.1 - 3 521 9056725
Gene Symbol: CT45A2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AY743710.1 - 3 622
Gene Symbol: CT45A5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007551.4 NP_001007552.2 3 622
NM_001172288.2 NP_001165759.2 3 639
XM_011531341.2 XP_011529643.1 3 504
GenBank mRNA BC028711.2 - 3 622 4828274
BC140735.1 - 3 529 9021603
BC140926.1 - 3 529 9056738
BC144437.1 - 3 529 9052959
Gene Symbol: CT45A6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017438.2 NP_001017438.2 3 622
XM_005262425.4 XP_005262482.2 3 542
XM_011531351.1 XP_011529653.1 3 473
GenBank mRNA AY743714.1 - 3 622
Gene Symbol: CT45A7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291543.1 NP_001278472.1 3 438
XM_003960082.4 XP_003960131.1 3 525
XM_011531229.2 XP_011529531.1 3 504
GenBank mRNA BU540102.1 - 3 441
Gene Symbol: CT45A8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BP371111.1 - 3 467

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0