This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OR2T35 SIRNA Assays ›
Gene Symbol:
OR2T35 OR2T2
Gene Name:
olfactory receptor family 2 subfamily T member 35
olfactory receptor family 2 subfamily T member 2
Gene Aliases:
-
OR1-43, OR2T2P
Chromosome Location:
Chr.1: 248638287 - 248639258 on Build GRCh38
Chromosome Location:
Chr.1: 248452798 - 248453772 on Build GRCh38
UniGene ID:
Hs.534846
Hs.554482
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID s53801

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: OR2T35
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001827.1 NP_001001827.1 1 800
Gene Symbol: OR2T2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004136.1 NP_001004136.1 1 803
GenBank mRNA BC136961.1 - 1 825 9020953
BC136962.1 - 1 825 9020954

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0