This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM166A SIRNA Assays ›
Gene Symbol:
FAM166A
Gene Name:
family with sequence similarity 166 member A
Gene Aliases:
HSD46
Chromosome Location:
Chr.9: 137243583 - 137251193 on Build GRCh38
UniGene ID:
Hs.522530
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s53655

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM166A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001710.2 NP_001001710.1 3 361
XM_011518694.1 XP_011516996.1 4 1346
XM_011518696.2 XP_011516998.1 4 1346
XM_011518697.1 XP_011516999.1 4 1346
XM_017014718.1 XP_016870207.1 3 1201
GenBank mRNA AK097419.1 - 3 339
AY604177.1 - 3 339
BC132916.1 - 3 306 40146938
BC132918.1 - 3 306 40146940
HY011475.1 - 2 443

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0