This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCNQ4 SIRNA Assays ›
Gene Symbol:
KCNQ4
Gene Name:
potassium voltage-gated channel subfamily Q member 4
Gene Aliases:
DFNA2, DFNA2A, KV7.4
Chromosome Location:
Chr.1: 40784012 - 40840457 on Build GRCh38
UniGene ID:
Hs.473058
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s536359

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KCNQ4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004700.3 NP_004691.2 11, 12 1697
NM_172163.2 NP_751895.1 10, 11 1535
XM_017002792.1 XP_016858281.1 8, 9 938
GenBank mRNA AF105202.1 - 11, 12 1697
AK074957.1 - 8, 9 934

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

potassium ion transport
sensory perception of sound
negative regulation of synaptic transmission, dopaminergic
regulation of ion transmembrane transport
inner ear morphogenesis
potassium ion transmembrane transport
delayed rectifier potassium channel activity
potassium channel activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0