This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MAGEL2 SIRNA Assays ›
Gene Symbol:
MAGEL2
Gene Name:
MAGE family member L2
Gene Aliases:
NDNL1, PWLS, SHFYNG, nM15
Chromosome Location:
Chr.15: 23643549 - 23647846 on Build GRCh38
UniGene ID:
Hs.141496
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s536277

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MAGEL2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019066.4 NP_061939.3 1 3551
GenBank mRNA AJ243531.1 - 1 1536
BC035839.1 - 1 1673
BC063834.1 - 1 1733
BC112257.1 - 1 1473
BC112259.1 - 1 1473

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
Arp2/3 complex-mediated actin nucleation
retrograde transport, endosome to Golgi
regulation of circadian rhythm
negative regulation of transcription, DNA-templated
rhythmic process
positive regulation of actin nucleation
protein K63-linked ubiquitination
ubiquitin-protein transferase activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0