This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FPR2 SIRNA Assays ›
Gene Symbol:
FPR2
Gene Name:
formyl peptide receptor 2
Gene Aliases:
ALXR, FMLP-R-II, FMLPX, FPR2A, FPRH1, FPRH2, FPRL1, HM63, LXA4R
Chromosome Location:
Chr.19: 51752026 - 51770526 on Build GRCh38
UniGene ID:
Hs.99855
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s5356

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FPR2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005738.1 NP_001005738.1 2 657
NM_001462.3 NP_001453.1 2 755
XM_006723120.3 XP_006723183.1 5 1488
XM_017026542.1 XP_016882031.1 4 1332
GenBank mRNA AF054013.1 - 1 612
AK290557.1 - 3 803
BC029125.2 - 1 626 5180065
BC071722.1 - 2 927 5212947
D10922.1 - 2 646
M76672.1 - 1 612
M84562.1 - 2 1384
M88107.1 - 2 651
U81501.1 - 1 632
X63819.1 - 2 765

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

movement of cell or subcellular component
chemotaxis
inflammatory response
cell adhesion
G-protein coupled receptor signaling pathway
G-protein coupled receptor activity
N-formyl peptide receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0