This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ARSE SIRNA Assays ›
Gene Symbol:
ARSE
Gene Name:
arylsulfatase E (chondrodysplasia punctata 1)
Gene Aliases:
ASE, CDPX, CDPX1, CDPXR
Chromosome Location:
Chr.X: 2934632 - 2968310 on Build GRCh38
UniGene ID:
Hs.386975
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s535040

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ARSE
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000047.2 NP_000038.2 10 1607
NM_001282628.1 NP_001269557.1 11 1893
NM_001282631.1 NP_001269560.1 9 1377
XM_005274518.2 XP_005274575.1 10 1534
XM_005274519.4 XP_005274576.1 10 1697
XM_005274521.4 XP_005274578.1 9 1490
XM_011545521.1 XP_011543823.1 8 1177
XM_017029525.1 XP_016885014.1 11 1824
XM_017029526.1 XP_016885015.1 9 1377
GenBank mRNA AK223183.1 - 10 1437
AK223199.1 - 10 1404
AK293648.1 - 10 1312
AK301004.1 - 11 1724
AK313093.1 - 10 1426
BC130438.1 - 10 1402 40146469
X83573.1 - 10 1424

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
glycosphingolipid metabolic process
post-translational protein modification
arylsulfatase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0