This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CUL7 SIRNA Assays ›
Gene Symbol:
CUL7
Gene Name:
cullin 7
Gene Aliases:
3M1, KIAA0076, dJ20C7.5
Chromosome Location:
Chr.6: 43037617 - 43053950 on Build GRCh38
UniGene ID:
Hs.520136
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s533990

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CUL7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006715285.1 XP_006715348.1 24, 25 4996
XM_011515019.2 XP_011513321.1 24, 25 4853
XM_011515020.2 XP_011513322.1 24, 25 4756
XM_011515021.1 XP_011513323.1 19, 20 2937
XM_017011533.1 XP_016867022.1 24, 25 4881
XM_017011535.1 XP_016867024.1 24, 25 4784
XM_017011536.1 XP_016867025.1 24, 25 5023
XM_017011538.1 XP_016867027.1 24, 25 4927
XM_017011539.1 XP_016867028.1 24, 25 4900

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
mitotic cytokinesis
vasculogenesis
epithelial to mesenchymal transition
placenta development
proteolysis
ubiquitin-dependent protein catabolic process
Golgi organization
regulation of mitotic nuclear division
viral process
protein ubiquitination
IRE1-mediated unfolded protein response
positive regulation of dendrite morphogenesis
protein binding
ubiquitin protein ligase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0