This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RBM10 SIRNA Assays ›
Gene Symbol:
RBM10
Gene Name:
RNA binding motif protein 10
Gene Aliases:
DXS8237E, GPATC9, GPATCH9, S1-1, TARPS, ZRANB5
Chromosome Location:
Chr.X: 47145196 - 47186815 on Build GRCh38
UniGene ID:
Hs.401509
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s533777

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RBM10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204466.1 NP_001191395.1 23 2929
NM_001204467.1 NP_001191396.1 24 3157
NM_001204468.1 NP_001191397.1 24 3191
NM_005676.4 NP_005667.2 24 3160
NM_152856.2 NP_690595.1 23 2926
XM_005272677.4 XP_005272734.1 24 3210
XM_005272678.4 XP_005272735.1 23 2744
XM_005272679.4 XP_005272736.1 23 2741
XM_006724563.1 XP_006724626.1 20 2377
XM_011543989.2 XP_011542291.1 16 2101
XM_017029884.1 XP_016885373.1 23 3080
XM_017029885.1 XP_016885374.1 23 2852
GenBank mRNA AK000962.1 - 10 1212
AK024839.1 - 7 768
AK025854.1 - 12 1533
AK225175.1 - 15 1832
AK292758.1 - 23 2880
AL137421.1 - 25 3049
BC003089.1 - 23 2865 3536058
BC004181.2 - 24 3113 2961094
BC008733.2 - 24 3145 3163064
BC024153.2 - 23 2775 3845258
BX537969.1 - 24 3142
D50912.1 - 24 3008
KU178411.1 - 22 2533
KU178412.1 - 23 2767
U35373.1 - 10 1144

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: D83948.
This siRNA also targets the following transcripts in Mouse: BC004674, BC157984, AK145991, AK122208, AK167666, and others.

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
mRNA processing
biological_process
RNA splicing
regulation of apoptotic process
nucleotide binding
protein binding
zinc ion binding
protein complex binding
identical protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0