This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FANCD2 SIRNA Assays ›
Gene Symbol:
FANCD2
Gene Name:
Fanconi anemia complementation group D2
Gene Aliases:
FA-D2, FA4, FACD, FAD, FAD2, FANCD
Chromosome Location:
Chr.3: 10026387 - 10101937 on Build GRCh38
UniGene ID:
Hs.208388
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s533672

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FANCD2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018115.2 NP_001018125.1 34 3485
NM_001319984.1 NP_001306913.1 34 3557
NM_033084.4 NP_149075.2 34 3485
GenBank mRNA AF230336.1 - 34 3428
AF340183.1 - 34 3428
AK074406.1 - 7 683
AL832427.1 - 18 1864

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

synapsis
gamete generation
response to gamma radiation
interstrand cross-link repair
protein binding
DNA polymerase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0