This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NBEAL2 SIRNA Assays ›
Gene Symbol:
NBEAL2
Gene Name:
neurobeachin like 2
Gene Aliases:
BDPLT4, GPS
Chromosome Location:
Chr.3: 46979683 - 47009704 on Build GRCh38
UniGene ID:
Hs.437043
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s532265

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NBEAL2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015175.2 NP_055990.1 32 5358
XM_006713072.3 XP_006713135.1 31 5274
XM_011533533.2 XP_011531835.1 32 5355
XM_017006010.1 XP_016861499.1 32 5355
XM_017006011.1 XP_016861500.1 32 5211
XM_017006012.1 XP_016861501.1 31 5274
XM_017006013.1 XP_016861502.1 32 5355
XM_017006014.1 XP_016861503.1 31 5130
XM_017006015.1 XP_016861504.1 30 4986
XM_017006016.1 XP_016861505.1 27 5932
XM_017006017.1 XP_016861506.1 9 1774
GenBank mRNA AB011112.3 - 18 3042
AK131104.1 - 32 5198
AY358455.1 - 2 308

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

platelet formation
phospholipid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header