This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMC1 SIRNA Assays ›
Gene Symbol:
TMC1
Gene Name:
transmembrane channel like 1
Gene Aliases:
DFNA36, DFNB11, DFNB7
Chromosome Location:
Chr.9: 72521801 - 72836351 on Build GRCh38
UniGene ID:
Hs.670211
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s531886

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMC1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138691.2 NP_619636.2 9 963
XM_017014256.1 XP_016869745.1 6 686
GenBank mRNA AF417578.1 - 9 963
AK307461.1 - 7 687
BC103955.1 - 4 325
BC103956.1 - 4 325
BC114380.1 - 4 325
BC141860.1 - 4 325

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

detection of mechanical stimulus involved in sensory perception of sound
vestibular reflex
auditory receptor cell development
calcium ion transmembrane transport
voltage-gated calcium channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0