This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PEX6 SIRNA Assays ›
Gene Symbol:
PEX6
Gene Name:
peroxisomal biogenesis factor 6
Gene Aliases:
HMLR2, PAF-2, PAF2, PBD4A, PDB4B, PXAAA1
Chromosome Location:
Chr.6: 42963873 - 42979243 on Build GRCh38
UniGene ID:
Hs.656425
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s531443

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PEX6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000287.3 NP_000278.3 9, 10 2047
NM_001316313.1 NP_001303242.1 9, 10 1783
NR_133009.1 9, 10 2047
XM_011514661.1 XP_011512963.1 8, 9 1963
GenBank mRNA AB051076.1 - 9, 10 1954
AB051077.1 - 9, 10 1690
AB051078.1 - 9, 10 1954
AB208906.1 - 9, 10 2007
AK314237.1 - 9, 10 1983
BC033487.1 - 9, 10 1962
BC048331.1 - 9, 10 1970 5189685
D83703.1 - 9, 10 2024

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein targeting to peroxisome
peroxisome organization
protein import into peroxisome matrix, translocation
protein stabilization
protein binding
ATP binding
protein C-terminus binding
ATPase activity
protein complex binding
ATPase activity, coupled

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0