This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM221B SIRNA Assays ›
Gene Symbol:
FAM221B
Gene Name:
family with sequence similarity 221 member B
Gene Aliases:
C9orf128
Chromosome Location:
Chr.9: 35817017 - 35829148 on Build GRCh38
UniGene ID:
Hs.128075
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s53058

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM221B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012446.3 NP_001012448.2 2 372
NR_052026.1 2 372
XM_006716768.3 XP_006716831.1 3 919
GenBank mRNA BC144503.1 - 3 350
BC146808.1 - 2 357 8860353
BX648702.1 - 2 372

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header