This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FLT3LG SIRNA Assays ›
Gene Symbol:
FLT3LG
Gene Name:
fms related tyrosine kinase 3 ligand
Gene Aliases:
FL, FLT3L
Chromosome Location:
Chr.19: 49474172 - 49487038 on Build GRCh38
UniGene ID:
Hs.428
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s5293

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FLT3LG
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204502.1 NP_001191431.1 5 430
NM_001204503.1 NP_001191432.1 4 399
NM_001278637.1 NP_001265566.1 5 458
NM_001278638.1 NP_001265567.1 5 447
NM_001459.3 NP_001450.2 5 430
XM_005258680.4 XP_005258737.3 4 420
XM_005258681.4 XP_005258738.3 4 420
XM_005258682.4 XP_005258739.3 4 421
XM_005258683.4 XP_005258740.3 4 420
XM_006723116.3 XP_006723179.2 4 421
XM_011526675.2 XP_011524977.1 5 466
XM_011526676.2 XP_011524978.1 5 431
XM_011526677.2 XP_011524979.1 4 420
XM_011526678.2 XP_011524980.1 5 466
XM_011526680.2 XP_011524982.1 4 420
XM_011526682.2 XP_011524984.1 4 437
XM_017026532.1 XP_016882021.1 4 421
XM_017026533.1 XP_016882022.1 4 356
XM_017026534.1 XP_016882023.1 4 329
XM_017026535.1 XP_016882024.1 5 453
GenBank mRNA AI792400.1 - 5 411
AK301136.1 - 4 391
BC006331.2 - 5 420
BC011914.2 - 5 420
BC028001.1 - 5 431 5198812
BC126293.1 - 5 387 8992009
BC136464.1 - 5 387 9020453
BC144129.1 - 5 387 9052649
DB490139.2 - 5 461
U03858.1 - 4 403
U04806.1 - 5 412

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
signal transduction
positive regulation of cell proliferation
lymphocyte differentiation
regulation of myeloid dendritic cell activation
positive regulation of natural killer cell differentiation
embryonic hemopoiesis
positive regulation of myoblast differentiation
positive regulation of cell cycle
positive regulation of transcription from RNA polymerase II promoter
homeostasis of number of cells within a tissue
positive regulation of cell proliferation in bone marrow
negative regulation of apoptotic process in bone marrow
positive regulation of osteoclast proliferation
positive regulation of myoblast fusion
receptor binding
cytokine activity
receptor tyrosine kinase binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0