This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HNF1A SIRNA Assays ›
Gene Symbol:
HNF1A
Gene Name:
HNF1 homeobox A
Gene Aliases:
HNF-1A, HNF1, IDDM20, LFB1, MODY3, TCF-1, TCF1
Chromosome Location:
Chr.12: 120978515 - 121002512 on Build GRCh38
UniGene ID:
Hs.654455
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s529261

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HNF1A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000545.6 NP_000536.5 3 780
NM_001306179.1 NP_001293108.1 3 780
XM_005253931.3 XP_005253988.1 3 833
GenBank mRNA AK298422.1 - 3 654
BC104908.1 - 3 599 8143911
BC104910.1 - 3 599 8143913
BC143483.1 - 3 599
HM116541.1 - 3 722
HM116542.1 - 3 722
HM116543.1 - 3 722
HM116547.1 - 3 722
HM116548.1 - 3 722
HM116552.1 - 4 339
HM116558.1 - 3 697
HM449088.1 - 3 602
HM449089.1 - 2 402
M57732.1 - 3 602
X71346.1 - 3 775
X71347.1 - 3 710

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
regulation of pronephros size
renal glucose absorption
glucose homeostasis
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
glucose import
positive regulation of transcription initiation from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
protein dimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm