This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OR5T1 SIRNA Assays ›
Gene Symbol:
OR5T1
Gene Name:
olfactory receptor family 5 subfamily T member 1
Gene Aliases:
OR11-179, OR5T1P
Chromosome Location:
Chr.11: 56275639 - 56276619 on Build GRCh38
UniGene ID:
Hs.553747
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s52703

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: OR5T1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004745.1 NP_001004745.1 1 838
GenBank mRNA BC136992.1 - 1 924 9020984
BC136993.1 - 1 924 9020985

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell-cell signaling
detection of chemical stimulus involved in sensory perception of smell
adenylate cyclase-activating adrenergic receptor signaling pathway
adrenergic receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0