This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM209A SIRNA Assays ›
Gene Symbol:
FAM209A FAM209B
Gene Name:
family with sequence similarity 209 member A
family with sequence similarity 209 member B
Gene Aliases:
C20orf106, dJ1153D9.3
C20orf107, dJ1153D9.4
Chromosome Location:
Chr.20: 56524729 - 56533590 on Build GRCh38
Chromosome Location:
Chr.20: 56527124 - 56555626 on Build GRCh38
UniGene ID:
Hs.504907
Hs.287759
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID s52269

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM209A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012971.3 NP_001012989.2 2 548
GenBank mRNA BC070179.1 - 2 510
BC107704.1 - 2 523 6614835
BX335293.2 - 2 548
Gene Symbol: FAM209B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013646.3 NP_001013668.2 2 564
XM_017027849.1 XP_016883338.1 4 1080
GenBank mRNA BC105792.1 - 3 529 6618487
DB517586.1 - 2 329

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0