This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KRTDAP SIRNA Assays ›
Gene Symbol:
KRTDAP
Gene Name:
keratinocyte differentiation associated protein
Gene Aliases:
KDAP, UNQ467
Chromosome Location:
Chr.19: 35487324 - 35490531 on Build GRCh38
UniGene ID:
Hs.112457
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s52161

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KRTDAP
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207392.2 NP_997275.1 3, 4 254
GenBank mRNA AJ293408.1 - 3, 4 215
AY359007.1 - 3, 4 177
BC130501.1 - 3, 4 254 40146532

Back To Top

More Information


Gene Ontology Categories:

Function(s)

cell differentiation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0