This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SBSN SIRNA Assays ›
Gene Symbol:
SBSN
Gene Name:
suprabasin
Gene Aliases:
UNQ698
Chromosome Location:
Chr.19: 35523367 - 35528371 on Build GRCh38
UniGene ID:
Hs.433484
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s51609

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SBSN
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166034.1 NP_001159506.1 1 384
NM_001166035.1 NP_001159507.1 1 384
NM_198538.3 NP_940940.1 1 384
XM_011526929.2 XP_011525231.1 1 404
XM_011526930.2 XP_011525232.1 1 360
XM_011526931.2 XP_011525233.1 1 404
GenBank mRNA AA055367.1 - 1 170
AK291305.1 - 1 344
AY358701.1 - 1 384
BC063640.1 - 1 326 4750640
BX648076.1 - 1 346

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0