This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KRT77 SIRNA Assays ›
Gene Symbol:
KRT77
Gene Name:
keratin 77
Gene Aliases:
K1B, KRT1B
Chromosome Location:
Chr.12: 52689612 - 52703524 on Build GRCh38
UniGene ID:
Hs.334989
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s51552

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KRT77
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175078.2 NP_778253.2 5 1052
XM_011538288.2 XP_011536590.1 5 814
GenBank mRNA AJ564104.1 - 5 1052
BC118598.1 - 6 1137 40108170
BC122558.1 - 5 1052 40108176

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
structural molecule activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0