This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM101B SIRNA Assays ›
Gene Symbol:
FAM101B
Gene Name:
family with sequence similarity 101 member B
Gene Aliases:
CFM1
Chromosome Location:
Chr.17: 439978 - 445940 on Build GRCh38
UniGene ID:
Hs.345588
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s51498

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM101B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182705.2 NP_874364.1 2 1636
GenBank mRNA AK093557.1 - 1 330

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

epithelial to mesenchymal transition
skeletal system morphogenesis
negative regulation of chondrocyte development
actin filament bundle organization
negative regulation of bone mineralization involved in bone maturation
filamin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0