This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CATSPER3 SIRNA Assays ›
Gene Symbol:
CATSPER3
Gene Name:
cation channel sperm associated 3
Gene Aliases:
CACRC
Chromosome Location:
Chr.5: 134967906 - 135011707 on Build GRCh38
UniGene ID:
Hs.631804
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s51290

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CATSPER3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178019.2 NP_821138.1 6, 7 1014
GenBank mRNA AF432876.1 - 6, 7 996
AK292427.1 - 6, 7 1013
AY156951.1 - 2, 3 216
BC101692.1 - 6, 7 974 8069198
BC110384.1 - 6, 7 1014 5166215

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

sodium ion transport
calcium ion transport
multicellular organism development
sperm motility
response to progesterone
ion transmembrane transport
regulation of ion transmembrane transport
sperm-egg recognition
sperm capacitation
calcium ion transmembrane transport
membrane depolarization during action potential
voltage-gated calcium channel activity
cation channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0