This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRSS48 SIRNA Assays ›
Gene Symbol:
PRSS48
Gene Name:
protease, serine 48
Gene Aliases:
ESSPL
Chromosome Location:
Chr.4: 151276482 - 151291856 on Build GRCh38
UniGene ID:
Hs.651266
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s51122

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PRSS48
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183375.2 NP_899231.2 2, 3 207
XM_011531921.2 XP_011530223.1 1, 2 653
GenBank mRNA BC121109.1 - 2, 3 211 40120369

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0