This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF517 SIRNA Assays ›
Gene Symbol:
ZNF517
Gene Name:
zinc finger protein 517
Gene Aliases:
-
Chromosome Location:
Chr.8: 144798876 - 144813033 on Build GRCh38
UniGene ID:
Hs.521942
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s50712

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF517
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317936.1 NP_001304865.1 5 515
NM_213605.2 NP_998770.2 5 536
NR_134284.1 5 536
XM_011517014.2 XP_011515316.1 5 1075
XM_011517015.2 XP_011515317.1 5 517
XM_011517017.2 XP_011515319.1 4 934
XM_017013384.1 XP_016868873.1 2 460
GenBank mRNA AK096527.1 - 6 1032
AK097278.1 - 3 1559
AK131440.1 - 5 536
BC126161.1 - 4 456 8991877
BC143922.1 - 3 329

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0