This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FABP7 SIRNA Assays ›
Gene Symbol:
FABP7
Gene Name:
fatty acid binding protein 7
Gene Aliases:
B-FABP, BLBP, FABPB, MRG
Chromosome Location:
Chr.6: 122749201 - 122784077 on Build GRCh38
UniGene ID:
Hs.26770
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s4984

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FABP7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319039.1 NP_001305968.1 2 444
NM_001319041.1 NP_001305970.1 2 444
NM_001319042.1 NP_001305971.1 2 185
NM_001446.4 NP_001437.1 2 444
GenBank mRNA AB208815.1 - 2 191
AI879148.1 - 2 487
AJ002962.1 - 2 200
AK289836.1 - 2 203
AK307272.1 - 2 204
AK311867.1 - 2 203
AL512688.1 - 2 160
BC012299.1 - 2 310 4707233
BI860090.1 - 2 418
BX461253.2 - 2 165
CR457057.1 - 2 124
D50373.1 - 2 176
D88648.1 - 2 175
KM111186.1 - 2 185
U51338.1 - 2 179
U81235.1 - 2 139

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
nervous system development
negative regulation of cell proliferation
triglyceride catabolic process
cell proliferation in forebrain
neurogenesis
epithelial cell proliferation
prepulse inhibition
transporter activity
lipid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0